Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSGALNACT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172170.1 | NP_001165641.1 | 1571 | 79 | ||
XM_028853368.1 | XP_028709201.1 | 408 | 79 | |||
GenBank mRNA | JU321709.1 | - | 1574 | 79 | ||
JU473204.1 | - | 1564 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |