Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSGALNACT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001172170.1 | NP_001165641.1 | 1661 | 81 | ||
XM_028853365.1 | XP_028709198.1 | 1443 | 81 | |||
XM_028853368.1 | XP_028709201.1 | 498 | 81 | |||
GenBank mRNA | JU321709.1 | - | 1664 | 81 | ||
JU473204.1 | - | 1654 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Macaca mulatta |