Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETNK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015151275.2 | XP_015006761.1 | 1676 | 111 | ||
GenBank mRNA | JV586543.1 | - | 830 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |