Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTTNBP2NL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257547.1 | NP_001244476.1 | 282 | 148 | ||
XM_015147066.2 | XP_015002552.1 | 487 | 148 | |||
GenBank mRNA | JU321676.1 | - | 282 | 148 | ||
JV044086.1 | - | 296 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |