Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MKKS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266509.2 | NP_001253438.1 | 1742 | 117 | ||
XM_015149380.2 | XP_015004866.1 | 1527 | 117 | |||
GenBank mRNA | JU322830.1 | - | 1718 | 117 | ||
JU473290.1 | - | 1742 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |