Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001117305.4 | XP_001117305.2 | 524 | 119 | ||
XM_015136314.2 | XP_014991800.1 | 853 | 119 | |||
GenBank mRNA | JU335059.1 | - | 491 | 119 | ||
JV510268.1 | - | 293 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |