Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASH3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261510.1 | NP_001248439.1 | 622 | 126 | ||
GenBank mRNA | JU472175.1 | - | 622 | 126 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |