Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093331.4 | XP_001093331.2 | 493 | 145 | ||
XM_015112590.2 | XP_014968076.2 | 288 | 145 | |||
GenBank mRNA | JU335076.1 | - | 540 | 145 | ||
JU476079.1 | - | 533 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |