Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ETAA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093331.4 | XP_001093331.2 | 604 | 150 | ||
XM_015112590.2 | XP_014968076.2 | 399 | 150 | |||
XM_028832438.1 | XP_028688271.1 | 258 | 150 | |||
GenBank mRNA | JU335076.1 | - | 651 | 150 | ||
JU476079.1 | - | 644 | 150 | |||
JV063789.1 | - | 115 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |