Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASNSD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001106941.4 | XP_001106941.2 | 191 | 145 | ||
GenBank mRNA | JU323128.1 | - | 154 | 145 | ||
JU472280.1 | - | 164 | 145 | |||
JV047437.1 | - | 55 | 145 | |||
JV703919.1 | - | 202 | 145 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |