Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPHK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257487.1 | NP_001244416.1 | 649 | 69 | ||
XM_015124381.2 | XP_014979867.1 | 1360 | 69 | |||
XM_015124382.2 | XP_014979868.1 | 1369 | 69 | |||
XM_015124383.2 | XP_014979869.1 | 592 | 69 | |||
XM_015124384.2 | XP_014979870.1 | 884 | 69 | |||
XM_028838491.1 | XP_028694324.1 | 805 | 69 | |||
GenBank mRNA | JU335508.1 | - | 649 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |