Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC698227 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015123918.2 | XP_014979404.1 | 3053 | 85 | ||
GenBank mRNA | FJ613343.1 | - | 256 | 85 | ||
FJ613344.1 | - | 256 | 85 | |||
JV513468.1 | - | 215 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta |