Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LZTFL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266443.1 | NP_001253372.1 | 267 | 132 | ||
GenBank mRNA | JU321609.1 | - | 247 | 132 | ||
JU472441.1 | - | 267 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |