Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLSCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265888.1 | NP_001252817.1 | 656 | 107 | ||
XM_015118456.2 | XP_014973942.1 | 1046 | 107 | |||
XM_015118457.2 | XP_014973943.1 | 1050 | 107 | |||
XM_028835783.1 | XP_028691616.1 | 1141 | 107 | |||
GenBank mRNA | JU321589.1 | - | 660 | 107 | ||
JV045213.1 | - | 656 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |