Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INTS12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260673.1 | NP_001247602.1 | 201 | 102 | ||
XM_015138908.2 | XP_014994394.1 | 540 | 102 | |||
XM_015138910.2 | XP_014994396.1 | 1047 | 102 | |||
XM_028847955.1 | XP_028703788.1 | 286 | 102 | |||
GenBank mRNA | JU327519.1 | - | 122 | 102 | ||
JU472078.1 | - | 201 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |