Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: APMAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266050.1 | NP_001252979.1 | 1105 | 99 | ||
XM_028827283.1 | XP_028683116.1 | 1546 | 99 | |||
XM_028827284.1 | XP_028683117.1 | 1337 | 99 | |||
XM_028827285.1 | XP_028683118.1 | 1267 | 99 | |||
GenBank mRNA | JU321400.1 | - | 1105 | 99 | ||
JV513791.1 | - | 1007 | 99 | |||
JV656575.1 | - | 1123 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |