Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AKIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194219.2 | NP_001181148.1 | 479 | 105 | ||
XM_001102761.3 | XP_001102761.1 | 325 | 105 | |||
XM_028832629.1 | XP_028688462.1 | 325 | 105 | |||
GenBank mRNA | JU335504.1 | - | 357 | 105 | ||
JU476442.1 | - | 397 | 105 | |||
JV047683.1 | - | 415 | 105 | |||
JV690157.1 | - | 421 | 105 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |