Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001193817.3 | NP_001180746.2 | 544 | 84 | ||
GenBank mRNA | JU322247.1 | - | 498 | 84 | ||
JU476528.1 | - | 495 | 84 |