Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMIGD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001302780.1 | NP_001289709.1 | 824 | 90 | ||
NM_001302781.1 | NP_001289710.1 | 217 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Macaca mulatta |