Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257565.1 | NP_001244494.1 | 1407 | 90 | ||
GenBank mRNA | JU335621.1 | - | 1407 | 90 | ||
JU476535.1 | - | 1426 | 90 | |||
JV527811.1 | - | 1437 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |