Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NCEH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001084048.4 | XP_001084048.2 | 832 | 148 | ||
XM_015131617.2 | XP_014987103.2 | 500 | 148 | |||
GenBank mRNA | JU326876.1 | - | 688 | 148 | ||
JV552433.1 | - | 716 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |