Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRMT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261618.1 | NP_001248547.1 | 766 | 146 | ||
XM_028850310.1 | XP_028706143.1 | 765 | 146 | |||
XM_028850311.1 | XP_028706144.1 | 768 | 146 | |||
GenBank mRNA | JU323425.1 | - | 745 | 146 | ||
JU473748.1 | - | 766 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |