Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANGPTL7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001103825.4 | XP_001103825.1 | 661 | 75 | ||
GenBank mRNA | JU478642.1 | - | 482 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |