Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260550.1 | NP_001247479.1 | 615 | 147 | ||
GenBank mRNA | JV043180.1 | - | 615 | 147 | ||
JV656237.1 | - | 432 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |