Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRAPPC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194511.2 | NP_001181440.1 | 416 | 144 | ||
XM_001111285.4 | XP_001111285.1 | 374 | 144 | |||
GenBank mRNA | JU320131.1 | - | 378 | 144 | ||
JU320132.1 | - | 377 | 144 | |||
JU473417.1 | - | 407 | 144 | |||
JV691979.1 | - | 400 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |