Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PCTP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001104194.4 | XP_001104194.2 | 438 | 153 | ||
XM_028836326.1 | XP_028692159.1 | 539 | 153 | |||
XM_028836327.1 | XP_028692160.1 | 539 | 153 | |||
XM_028836328.1 | XP_028692161.1 | 567 | 153 | |||
XM_028836329.1 | XP_028692162.1 | 539 | 153 | |||
XM_028836330.1 | XP_028692163.1 | 539 | 153 | |||
XM_028836331.1 | XP_028692164.1 | 539 | 153 | |||
XM_028836332.1 | XP_028692165.1 | 539 | 153 | |||
XM_028836333.1 | XP_028692166.1 | 438 | 153 | |||
XM_028836334.1 | XP_028692167.1 | 438 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |