Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KMT2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015135341.2 | XP_014990827.2 | 9329 | 146 | ||
XM_015135342.2 | XP_014990828.2 | 8720 | 146 | |||
XM_028846497.1 | XP_028702330.1 | 9584 | 146 | |||
XM_028846498.1 | XP_028702331.1 | 9526 | 146 | |||
XM_028846499.1 | XP_028702332.1 | 9526 | 146 | |||
XM_028846500.1 | XP_028702333.1 | 9526 | 146 | |||
XM_028846501.1 | XP_028702334.1 | 9526 | 146 | |||
XM_028846502.1 | XP_028702335.1 | 9493 | 146 | |||
XM_028846503.1 | XP_028702336.1 | 9526 | 146 | |||
XM_028846504.1 | XP_028702337.1 | 9811 | 146 | |||
XM_028846505.1 | XP_028702338.1 | 9447 | 146 | |||
XM_028846506.1 | XP_028702339.1 | 9245 | 146 | |||
XM_028846507.1 | XP_028702340.1 | 9670 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |