Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC715304 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001105454.4 | XP_001105454.2 | 1167 | 99 | ||
XM_015135992.2 | XP_014991478.1 | 185 | 99 | |||
GenBank mRNA | KU253647.1 | - | 970 | 99 | ||
KU253649.1 | - | 1348 | 99 | |||
KU253650.1 | - | 281 | 99 | |||
KU253651.1 | - | 348 | 99 | |||
KU253653.1 | - | 278 | 99 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |