Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194242.1 | NP_001181171.1 | 279 | 158 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |