Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028835225.1 | XP_028691058.1 | 1754 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Macaca mulatta |