Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CUZD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148386.2 | XP_015003872.1 | 939 | 161 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |