Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SNX16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145621.2 | XP_015001107.1 | 1125 | 87 | ||
XM_015145622.2 | XP_015001108.1 | 1026 | 87 | |||
GenBank mRNA | JU322696.1 | - | 1050 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta |