Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCTA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260695.1 | NP_001247624.1 | 287 | 117 | ||
GenBank mRNA | JU336123.1 | - | 344 | 117 | ||
JU336124.1 | - | 344 | 117 | |||
JU336125.1 | - | 344 | 117 | |||
JU476986.1 | - | 287 | 117 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |