Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TTC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015112692.2 | XP_014968178.1 | 311 | 155 | ||
XM_015112694.2 | XP_014968180.1 | 311 | 155 | |||
GenBank mRNA | JU325245.1 | - | 109 | 155 | ||
JU472254.1 | - | 759 | 155 | |||
JV636321.1 | - | 1712 | 155 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |