Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MS4A5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086601.4 | XP_001086601.1 | 287 | 170 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |