Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194825.1 | NP_001181754.1 | 188 | 60 | ||
XM_015123314.2 | XP_014978800.1 | 287 | 60 | |||
XM_028838552.1 | XP_028694385.1 | 311 | 60 | |||
GenBank mRNA | JU336339.1 | - | 190 | 60 | ||
JU477162.1 | - | 250 | 60 | |||
JV045587.1 | - | 173 | 60 | |||
JV529003.1 | - | 212 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |