Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC28B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001102480.4 | XP_001102480.2 | 863 | 139 | ||
GenBank mRNA | JU336354.1 | - | 612 | 139 | ||
JU477174.1 | - | 616 | 139 | |||
JV048230.1 | - | 601 | 139 | |||
JV638399.1 | - | 657 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |