Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122718.2 | XP_014978204.1 | 496 | 84 | ||
XM_015122719.2 | XP_014978205.1 | 371 | 84 | |||
XM_015122720.2 | XP_014978206.1 | 527 | 84 | |||
GenBank mRNA | JU336384.1 | - | 214 | 84 | ||
JV048252.1 | - | 180 | 84 | |||
JV696363.1 | - | 182 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Macaca mulatta |