Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CERS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015122718.2 | XP_014978204.1 | 613 | 143 | ||
XM_015122719.2 | XP_014978205.1 | 488 | 143 | |||
XM_015122720.2 | XP_014978206.1 | 644 | 143 | |||
GenBank mRNA | JU336384.1 | - | 331 | 143 | ||
JV048252.1 | - | 297 | 143 | |||
JV696363.1 | - | 299 | 143 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |