Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OGFRL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015136683.2 | XP_014992169.1 | 1131 | 116 | ||
XM_015136684.2 | XP_014992170.1 | 1128 | 116 | |||
XM_015136685.2 | XP_014992171.1 | 406 | 116 | |||
GenBank mRNA | JU323158.1 | - | 605 | 116 | ||
JV723371.1 | - | 657 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |