Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GAL3ST4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260777.1 | NP_001247706.1 | 372 | 68 | ||
GenBank mRNA | JU473256.1 | - | 350 | 68 | ||
JV045325.1 | - | 372 | 68 | |||
JV636378.1 | - | 361 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta |