Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: QTRT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265882.2 | NP_001252811.1 | 567 | 107 | ||
XM_015129814.2 | XP_014985300.2 | 701 | 107 | |||
GenBank mRNA | JU325373.1 | - | 495 | 107 | ||
JU325374.1 | - | 495 | 107 | |||
JV045284.1 | - | 568 | 107 | |||
JV637350.1 | - | 574 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |