Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015131644.2 | XP_014987130.2 | 779 | 144 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |