Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CAMKMT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001112368.4 | XP_001112368.3 | 893 | 116 | ||
GenBank mRNA | JU326690.1 | - | 535 | 116 | ||
JV723624.1 | - | 676 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |