Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KDM8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015125827.2 | XP_014981313.2 | 1040 | 110 | ||
XM_015125828.2 | XP_014981314.2 | 1030 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |