Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CSRNP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257746.1 | NP_001244675.1 | 649 | 147 | ||
XM_015110246.1 | XP_014965732.1 | 788 | 147 | |||
XM_015110248.2 | XP_014965734.1 | 603 | 147 | |||
GenBank mRNA | JU322251.1 | - | 649 | 147 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |