Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIKE1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261494.1 | NP_001248423.1 | 580 | 128 | ||
XM_015147217.2 | XP_015002703.1 | 619 | 128 | |||
GenBank mRNA | JU336479.1 | - | 577 | 128 | ||
JU477288.1 | - | 580 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Macaca mulatta |