Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROSER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087439.4 | XP_001087439.2 | 1491 | 68 | ||
XM_015120959.2 | XP_014976445.2 | 823 | 68 | |||
GenBank mRNA | JU477301.1 | - | 1479 | 68 | ||
JU477302.1 | - | 1479 | 68 | |||
JV048305.1 | - | 816 | 68 | |||
JV724005.1 | - | 1141 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Macaca mulatta |