Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266726.2 | NP_001253655.1 | 1703 | 129 | ||
XM_015147269.2 | XP_015002755.1 | 1215 | 129 | |||
GenBank mRNA | JU336496.1 | - | 1675 | 129 | ||
JV045067.1 | - | 1706 | 129 | |||
JV637095.1 | - | 1689 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Macaca mulatta |