Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: REEP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266282.1 | NP_001253211.1 | 635 | 79 | ||
GenBank mRNA | JU477316.1 | - | 731 | 79 | ||
JV048313.1 | - | 730 | 79 | |||
JV636389.1 | - | 635 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Macaca mulatta |